Learn More
Enzo Life Sciences DNA topoisomerase II alpha (pThr1342) monoclonal antibody (3D4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC220E
Encompass_Preferred
Clone: 3D4. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of human Topoisomerase IIα phosphorylated at Tyr1342. Species reactivity: Human. Applications: ICC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7245