Learn More
ABclonal Technology DNA topoisomerase I (TOP1) Rabbit pAb
SDP
Supplier:  ABclonal Technology A125241000UL
Encompass_Preferred
This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
Catalog No. 50-001-14783