Learn More
ATCC Genomic DNA, Vaccinia virus strain IHD - frozen genomic DNA, 100 μL vial for PCR and molecular biology
Supplier:  ATCC VR156D
Encompass
Genomic DNA isolated from Vaccinia virus strain IHD supplied as a frozen aliquot for molecular biology applications, including PCR and sequencing. Intended for laboratory research use only; not for diagnostic or therapeutic purposes.
  • Supplied as a frozen 100 μL aliquot.
  • Intended for PCR, sequencing, and other molecular biology workflows.
  • Store at -70°C or colder; shipped frozen.
  • Lot-specific concentration information available on request.
  • Derived from the IHD strain of Vaccinia virus; handle following institutional biosafety procedures.
  • Biosafety level 1 advisory; follow local regulations and containment practices.
Catalog No. NC3038648