Learn More
NANOMICRO TECHNOLOGIES INC NanoChrom Technologies, DNACore SEC-2000 LC Analytical Column, 3um, 4.6 x 300 mm, Stainless Steel
Supplier:  NANOMICRO TECHNOLOGIES INC D20303020004630S
Encompass_Preferred
DNACore SEC is a family of high performance, size exclusion chromatography columns designed for separating plasmids, circle RNA, RV, AV, lentivirus and related aggregates and fragments. The column technology involves the creation of a neutral hydrophilic layer on the surface of specially designed, wide-pore, monodispersed silica particles, combined with well-established column packing processes.
Catalog No. NC3587988