Learn More
Genscript Corporation DNAM-1/CD226Biotin His Av
Supplier:  Genscript Corporation Z04442500
Encompass
DNAX accessory molecule-1 (DNAM-1) also known as CD226 is a 65 kDa type I transmembrane glycoprotein in the immunoglobulin superfamily DNAM-1 mediates cellular adhesion to other cells bearing its ligands CD112 and CD155 and cross-linking DNAM-1 with antibodies causes cellular activation Furthermore DNAM-1 can interact with PVR and PVRL2
Catalog No. 50-000-58793