Learn More
Cayman Chemical DNARNA OxIdatIve Damage Mon
SDP
Supplier:  Cayman Chemical 20094100 UG
Encompass_Preferred
Immunogen: 8-Hydroxy-2-deoxyguanosine • Host: Mouse • Cross Reactivity: (+) 8-hydroxy-2-deoxyguanosine, 8-hydroxyguanosine • Applications: Affinity purification, ELISA
Catalog No. 50-298-2864