Learn More
LUNA NANOTECH INC Dnase I, Rnase-Free, 25 X 550U
Supplier:  LUNA NANOTECH INC GDN2550
Encompass_Preferred
DNase I, RNase-Free, Lyophilized Powder, 25 x 550U
DNase I (Deoxyribonuclease I) is an endonuclease enzyme that can degrade DNA by cleaving phosphodiester linkages in the DNA backbone. DNase I is commonly used in RNA purification protocols to remove contaminating DNA.
This DNase I enzyme is RNase-free, supplied as lyophilized powder. Each tube contains DNAse I enzyme with a total activity of 550U (units).
Catalog No. 50-256-6991