Learn More
Aldevron Dnase I, 500ug
Supplier:  Aldevron 9136500MCG
Encompass
500ug dispense of Dnase I. DNase I nonspecifically cleaves single- or double-stranded DNA, producing di-, tri- and oligonucleotide products. DNase I is free of detectable ribonuclease activity.
Catalog No. 50-307-3885