Learn More
AdipoGen DNER ED h rec
SDP
Supplier:  AdipoGen AG40A0137YC010
Encompass_Preferred
Protein. Signal peptide and extracellular domain of human DNER aa 1-640 are fused at the C-terminus with a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.02% CHAPS. Purity >90% SDS-PAGE. DNER is an activator of the Notch1 pathway. It mediates neuron-glia interaction during astrocytogenesis.
Catalog No. 50-256-2979