Learn More
Jackson Immuno Research Labs Alexa Fluor 594 AffiniPure F(ab')₂ Fragment Donkey Anti-Mouse IgG (H+L), .3mg
SDP
Supplier:  Jackson Immuno Research Labs 715586150
Encompass_Preferred
Alexa Fluor 594-AffiniPure F(ab')2 Fragment Donkey Anti-Mouse IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Rb,Shp Sr Prot)
Catalog No. NC0529642