Learn More
Medchemexpress LLC DNP-PEG4-DBCO | 2149585-17-9 | 97.0% | C35H39N5O10 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140619
Encompass_Preferred
DNP-PEG4-DBCO is a PEG-based PROTAC linker employed in the synthesis of PROTACs. This click chemistry reagent contains a DBCO group, allowing it to undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Used for PROTACs synthesis
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC reactions
  • Enables targeted protein degradation
Catalog No. 50-003-55955