Learn More
Medchemexpress LLC DNP-PEG4-NHS ester | 858126-78-0 | 96.81% | C21H28N4O12 | 100 MG

Supplier: Medchemexpress LLC HY140614100MG
DNP-PEG4-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- For research use only
- Stable for 3 years in pure form at -20°C
- Stable for 6 months in solvent at -80°C
- Stable for 1 month in solvent at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.