Learn More
Medchemexpress LLC DNP-PEG4-NHS ester | 858126-78-0 | 96.81% | C21H28N4O12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14061450MG
Encompass_Preferred
DNP-PEG4-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This compound facilitates the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system. PROTACs feature two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other for the protein of interest.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57019