Learn More
Cayman Chemical Dnsyl ChlorIde 1g
SDP
Supplier:  Cayman Chemical 274581 G
Encompass_Preferred
A reactive probe for derivatization of primary amines, including those on amino acids, peptides, and polyamines, for detection by HPLC; has been used for the derivatization of phenol-containing compounds, such as steroids, for detection by MS/MS; a fluorescent probe for proteins and enzymes with ex/em = 340/535 nm, respectively, in acetone
Catalog No. 50-294-1824