Learn More
Medchemexpress LLC Do34 | 1848233-58-8 | 99.3% | 531.53 | C26H28F3N5O4 | 1 ML

Supplier: Medchemexpress LLC HY11777110MM 1ML
DO34 is a potent, selective, and centrally active diacylglycerol lipase (DAGL) inhibitor used in preclinical research to reduce 2-AG levels and modulate endocannabinoid signaling. It is supplied as a DMSO solution or as a solid for experimental flexibility.
- Highly potent DAGL inhibitor (IC50 ~6 nM for DAGLα; 3-8 nM for DAGLβ).
- Reduces brain 2-AG levels in a dose- and time-dependent manner.
- Available as a 10 mM solution in DMSO (1 mL) and as solid quantities.
- High purity suitable for research (99.3% by HPLC).
- Molecular weight 531.53 g·mol⁻¹; formula C26H28F3N5O4.
- Soluble in DMSO; manufacturer provides in vivo formulation protocols.
- Store powder at -20°C; store solutions at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.