Learn More
Medchemexpress LLC DO34 analog | 2098969-71-0 | 98.4% | 531.53 g/mol | C26H28F3N5O4 | 1 ML

Supplier: Medchemexpress LLC HY117771A10MM 1ML
DO34 analog is a triazole-based diacylglycerol lipase (DAGLα) inhibitor (patent WO2017096315, compound 100) intended for biochemical and cell-based research. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO, providing high purity and convenient handling for in vitro studies.
- High purity: 98.42%.
- Molecular weight: 531.53 g/mol.
- Chemical formula: C26H28F3N5O4.
- Physical form: Light yellow to yellow solid; also supplied as a solution.
- Solubility: DMSO ≥ 105 mg/mL (≈197.5 mM).
- Storage: Powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).
- Available formats: Solids (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and 10 mM 1 mL solution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.