Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DO3A tert-Butyl ester | 122555-91-3 | 98.0% | 10 G
SDP

Supplier:  Medchemexpress LLC HY1269210G

Encompass_Preferred

DO3A tert-Butyl ester, also known as DOTA tert-Butyl ester, is a cyclic tosamide benzyl derivative for research applications. It shows cytotoxicity in cancer cell lines (A375, A2780, HT29, MCF-7, FaDu, NIH 3T3) with IC50s from 8.7 to 17.3 μM. Its molecular weight is 514.70, formula C26H50N4O6, and it appears as a white to yellow solid.

  • For research use only
  • Exhibits cytotoxicity in several cancer cell lines
  • White to yellow solid appearance
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 2 years, -20°C for 1 year
  • Shipped at room temperature if less than 2 weeks
  • Keep container tightly sealed in cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition

Catalog No. 50-003-41547


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.