Learn More
Cayman Chemical DobutamInhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 155825 MG
Encompass_Preferred
A β-adrenergic receptor agonist with potent positive inotropic effects in vivo; strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions; strongly increases cardiac contractility
Catalog No. 50-297-0845