Learn More
Enzo Life Sciences Docetaxel (5 mg). CAS: 114977-28-5
SDP
Supplier:  Enzo Life Sciences BMLT1290005
Encompass_Preferred
Docetaxel is taxane-type antineoplastic agent structurally similar to taxol. It reversibly binds to polymerized tubulin inhibiting depolymerization. It induces apoptosis in a large number of tumor cells and is a clinically important anticancer agent. Alternative name: Taxotere. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2218