Learn More
Medchemexpress LLC Docetaxel Trihydrate | 148408-66-6 | 99.91% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0011A10MM/1ML
Encompass_Preferred
Docetaxel Trihydrate is an antineoplastic agent that inhibits microtubule depolymerization. It is a semisynthetic analog of taxol and works by attenuating the effects of bcl-2 and bcl-xL gene expression. This leads to the arrest of the cell cycle at G2/M and subsequent cell apoptosis. It is intended for research use only.
  • Inhibits microtubule depolymerization
  • Semisynthetic analog of taxol
  • Attenuates effects of bcl-2 and bcl-xL gene expression
  • Arrests the cell cycle at G2/M
  • Leads to cell apoptosis
Catalog No. 50-004-32206