Learn More
Cayman Chemical Docetaxelhydrate 250mg
SDP
Supplier:  Cayman Chemical 11637250 MG
Encompass_Preferred
A microtubule-stabilizing agent; promotes microtubule assembly in a cell-free assay at 20 µM; is cytotoxic to J774.2 cells (EC50 = 0.05 µM); inhibits tumor growth in several cancer models, including B16 murine melanoma and P388 murine leukemia models
Catalog No. 50-295-5731