Learn More
Cayman Chemical Docosahexaenoyl GlycIn 10mg
SDP
Supplier:  Cayman Chemical 900032810 MG
Encompass_Preferred
Consists of docosahexaenoic acid, an ω-3 PUFA, with glycine attached at its carboxy terminus
Catalog No. 50-289-4695