Learn More
Medchemexpress LLC Docosahexaenoyl Serinol | 1638355-62-0 | 401.58 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1609271MG
Encompass_Preferred
Docosahexaenoyl Serinol (Compound 1) inhibits proliferation of lung cancer cells A549, with a 77.4% inhibition rate at 3 μM.
  • Inhibits proliferation of lung cancer cells
Catalog No. 50-003-94275