Learn More
Medchemexpress LLC Docosane | 629-97-0 | MFCD00009348 | 98.0% | 310.60 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN992910MM/1ML
Encompass_Preferred
Docosane is a straight-chain alkane that can be utilized to synthesize structural composites with thermal energy storage/release capabilities. It can also be employed in the preparation of thermosensitive antipyretic analgesics.
  • Straight-chain alkane
  • Utilized to synthesize structural composites with thermal energy storage/release capabilities
  • Employed in the preparation of thermosensitive antipyretic analgesics
Catalog No. 50-004-26508