Learn More
Medchemexpress LLC Docosanedioic acid | 505-56-6 | MFCD00002806 | 97.0% | 370.57 g/mol | C22H42O4 | 5 G

Supplier: Medchemexpress LLC HYW0349185G
Docosanedioic acid is a saturated dicarboxylic acid used as a non-cleavable linker for antibody-drug conjugate (ADC) synthesis and as an alkyl chain linker for proteolysis-targeting chimera (PROTAC) construction. Supplied as a powder for research use.
- Used as a non-cleavable ADC linker.
- Used as an alkyl chain PROTAC linker.
- Molecular formula C22H42O4.
- Molecular weight 370.57 g/mol.
- Physical appearance powder.
- Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Solubility: DMSO 50 mg/mL (ultrasonic recommended); in vivo formulation 10% DMSO/90% corn oil, ≥5 mg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.