Learn More
Medchemexpress LLC Docosanoic acid | 112-85-6 | 99.40% | 340.59 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW013049
Encompass_Preferred
Docosanoic acid, also known as Behenic acid, is a long-chain saturated fatty acid. It has been observed to inhibit the double-stranded DNA (dsDNA) binding activity of the p53 DNA binding domain, with a Kd of 12 nM. This compound has low bioavailability and can lead to an increase in cholesterol levels in humans.
  • Long-chain saturated fatty acid.
  • Inhibits dsDNA binding activity of p53 DNA binding domain (Kd = 12 nM).
  • 50% inhibition of wild-type p53 DBD observed at 0.41 nmol.
  • Inhibitory effect on R248A mutant p53 DBD is similar to wild-type.
  • In rats, dietary behenic acid (1.7%) significantly reduces visceral fat deposition, leading to lower adipose and hepatic tissue weight.
  • Low bioavailability, leading to reduced absorption and a decrease in body weight and visceral fat deposition in rats.
Catalog No. 50-004-53460