Learn More
Medchemexpress LLC Docosanoic acid-d3 (Behenic acid-d3) | 1193721-67-3 | 99.8% | 343.60 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW013049S310MG
Encompass_Preferred
Docosanoic acid-d3 (Behenic acid-d3) is a deuterium-labeled form of Docosanoic acid. This long-chain saturated fatty acid is known to inhibit the double-stranded DNA (dsDNA) binding activity of the p53 DNA binding domain, with a Kd of 12 nM. Docosanoic acid has low bioavailability and can increase cholesterol in humans. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitation during drug development. Deuteration can also affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Inhibits double-stranded DNA (dsDNA) binding activity of p53 DNA binding domain
  • Useful as a tracer or internal standard for quantitative analysis
  • Can affect pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-004-52176