Learn More
Selleck Chemical LLC Docosanol 50mg 661-19-8
SDP
Supplier:  Selleck Chemical LLC S163750MG
Encompass_Preferred
Docosanol is a saturated fatty alcoholused traditionally as an emollient, emulsifier, and thickener in cosmetics, nutritional supplement. (ED50=2.5 mg/mL for plaque inhibition, and ED50=1.7 mg/mL for inhibition of virus production) *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-679-9