Learn More
Cayman Chemical DocosapentaeNIc ACD mthyl 50mg
SDP
Supplier:  Cayman Chemical 900187050 MG
Encompass_Preferred
An esterified form of docosapentaenoic acid; has been used as a reference standard in the quantification of fatty acids in microalgal and fish oils
Catalog No. 50-289-3454