Learn More
Cayman Chemical DocosapentaenoIc AcId 10mg
SDP
Supplier:  Cayman Chemical 9016510 MG
Encompass_Preferred
Docosapentaenoic acid is an ω-3 fatty acid found in fish oils. It is a minor constituent of the total serum unsaturated fatty acids in humans, ranging from 0.1 to 1%, and increases on dietary supplementation.{1936}
Catalog No. 50-289-5764