Learn More
Cayman Chemical Docosapentaenoic Acid, 24880-45-3, 10 mg
SDP
Supplier:  Cayman Chemical 9016510
Encompass_Preferred
Molecular Formula C22H34O2, Purity ≥98%, Formula Weight 330.5, Synonyms: DPA; Docosapentaenoic acid is an -3 fatty acid found in fish oils. It is a minor constituent of the total serum unsaturated fatty acids in humans, ranging from 0.1 to 1%, and increases on dietary supplementation.{1936}
Catalog No. NC0369108