Learn More
Cayman Chemical Docosatetraenoyl Ethanolamide
SDP
Supplier:  Cayman Chemical 903855 MG
Encompass_Preferred
An endocannabinoid containing docosatetraenoic acid in place of the arachidonate moiety of AEA; acts on CB1 receptors with a potency and efficacy similar to that of AEA but its specific role and relative importance as a cannabinergic neurotransmitter have not been elucidated
Catalog No. 50-289-4863