Learn More
Cayman Chemical DocosnoIc AcId methyl ester 5g
SDP
Supplier:  Cayman Chemical 268665 G
Encompass_Preferred
An ester form of docosanoic acid; has been found in soybean, corn, sunflower, and canola oils, as well as beef fat, used in the production of biodiesel
Catalog No. 50-293-2783