Learn More
Medchemexpress LLC Docusate Sodium | 577-11-7 | 98.0% | 1 ML
Supplier: Medchemexpress LLC HYB126810MM/1ML
Docusate Sodium is a key ingredient in stool softeners. It acts as a sulfated surfactant that can inactivate viral pathogens by disrupting viral envelopes and/or denaturing/disassociating proteins. It has shown effectiveness in vitro against both wild-type and drug-resistant strains of HSV type 1 and 2. Additionally, Docusate Sodium is identified as an obesogen; developmental exposure to it has been linked to increased adult adiposity, inflammation, metabolic disorder, and dyslipidemia in offspring fed a standard diet in mice.
- One of the main components in stool softeners.
- May inactivate viral pathogens by disrupting viral envelopes and/or denaturing/disassociating proteins.
- Effective in vitro against wild type and drug-resistant strains of HSV type 1 and 2.