Learn More
Medchemexpress LLC Dodecaethylene glycol | 6790-09-6 | 99.9% | C24H50O13 | 10 G
SDP
Supplier:  Medchemexpress LLC HY14123310G
Encompass_Preferred
Dodecaethylene glycol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Solid appearance
  • White to off-white color
Catalog No. 50-003-55306