Learn More
Medchemexpress LLC Dodecane-1,12-diamine | 2783-17-7 | MFCD00008155 | 97.0% | 200.36 | C12H28N2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01488350G
Encompass_Preferred
1,12-Dodecanediamine is an aliphatic alpha,omega-diamine (dodecane-1,12-diamine; C12H28N2; MW 200.36; CAS 2783-17-7) used in ion channel research and as a chemical building block in polymer and surfactant synthesis. It is supplied as a white to off-white solid with documented purity for research applications.
  • Acts as a reversible Maxi calcium-activated potassium channel blocker for ion channel studies.
  • White to off-white solid form facilitates handling and storage.
  • Offered in laboratory pack sizes suitable for small-scale research.
  • High-purity variants available to support analytical and biological assays.
  • Soluble in common organic solvents for synthesis and formulation workflows.
Catalog No. 50-003-02965