Learn More
Medchemexpress LLC Dodecanedioic acid | 693-23-2 | ≥98.0% | 230.30 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01224150G
Encompass_Preferred
Dodecanedioic acid is an orally active, straight-chain dicarboxylic acid that serves as an alternative energy substrate between lipids and carbohydrates. It is rapidly oxidized during exercise, which helps reduce muscle fatigue without stimulating insulin secretion. This compound also exhibits blood glucose-lowering activity in non-insulin-dependent diabetes mellitus.
  • Orally active straight-chain dicarboxylic acid
  • Alternative energy substrate
  • Rapidly oxidized during exercise
  • Helps reduce muscle fatigue
  • Does not stimulate insulin secretion
  • Exhibits blood glucose-lowering activity in non-insulin-dependent diabetes mellitus
Catalog No. 50-004-53803