Learn More
Medchemexpress LLC Dodecanoylcarnitine ((-)-lauroylcarnitine) | 25518-54-1 | 99.8% | 343.50 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11316610MG
Encompass_Preferred
Dodecanoylcarnitine ((-)-Lauroylcarnitine) is a medium long-chain acylcarnitine, an intermediate in key energy metabolic pathways involving fatty acid β-oxidation and amino acid catabolism. Abnormal levels indicate mitochondrial fuel metabolism disturbance and are associated with type II diabetes, pre-diabetes, fatty acid oxidation disorders, and celiac disease. It also shows high sensitivities and specificities in predicting asthma.
  • Intermediate in fatty acid β-oxidation and amino acid catabolism.
  • Levels indicate mitochondrial fuel metabolism disturbances.
  • Associated with type II diabetes and pre-diabetes status.
  • Linked to fatty acid oxidation disorders and celiac disease.
  • Useful in predicting asthma.
  • Potential biomarker component for Yin-deficiency-heat syndrome.
Catalog No. 50-004-36364