Learn More
Medchemexpress LLC Dodecyl azide | 13733-78-3 | MFCD18351855 | 95.0% | 211.35 g/mol | C12H25N3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW590543100MG
Encompass_Preferred
Dodecyl azide (1-azidododecane) is a linear alkyl azide with a terminal azide (N3) group attached to a 12-carbon chain. It is used as an azide partner in copper-catalyzed and strain-promoted azide-alkyne cycloadditions (click chemistry) for labeling, conjugation, and synthetic transformations where a hydrophobic spacer is required. The compound has CAS 13733-78-3, formula C12H25N3, and molecular weight 211.35 g/mol.
  • Terminal azide functional group for click chemistry.
  • 12-carbon hydrophobic spacer suitable for conjugation and labeling.
  • Compatible with alkynes, BCN, and DBCO reagents.
  • Provided as a small-molecule research reagent for synthetic and bioconjugation use.
  • Store solid at -20°C for long-term stability; store solutions at lower temperatures.
Catalog No. 50-002-84798