Learn More
Medchemexpress LLC Dodecyl β-D-glucopyranoside | 59122-55-3 | MFCD00063298 | 98.0% | 348.47 | C18H36O6 | 5 G

Supplier: Medchemexpress LLC HYW1426925G
Dodecyl β-D-glucopyranoside is a non-ionic surfactant used to solubilize and stabilize membrane proteins and membrane-associated complexes in biochemical research. It is supplied as a high-purity, white to off-white solid with specified storage conditions to preserve chemical integrity.
- Non-ionic detergent suitable for membrane protein solubilization.
- High purity: 98.0%.
- Molecular formula C18H36O6 and molecular weight 348.47.
- White to off-white solid appearance.
- Optical rotation: -24.47° (C = 0.01 g/mL, MeOH, 20°C, 589 nm).
- Recommended storage: -20°C sealed; in solvent stability: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.