Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dodecyl dihydrogen phosphate | 2627-35-2 | MFCD00013946 | >95.0% | 266.32 g/mol | C12H27O4P | 100MG
SDP

Supplier:  Medchemexpress LLC HY125802100MG

Encompass_Preferred

Monolauryl phosphate is a phosphoric ester (dodecyl dihydrogen phosphate) used as a biologically active reagent in biochemical research. It is supplied for research purposes only and is not intended for human or clinical use.

  • Phosphoric ester reagent used in biochemical research.
  • CAS 2627-35-2.
  • Chemical formula C12H27O4P and molecular weight 266.32 g/mol.
  • Purity >95.0% (qNMR) as reported by a supplier.
  • Available in small laboratory quantities (50 mg-1 g).
  • Typical appearance: white to light yellow solid.

Catalog No. 50-002-38006


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.