Learn More
Medchemexpress LLC Dodecyl gallate | 1166-52-5 | MFCD00002195 | 99.8% | 338.4 g/mol | C19H30O5 | 50MG
SDP
Supplier:  Medchemexpress LLC HY124082R50MG
Encompass_Preferred
Dodecyl gallate (lauryl gallate) analytical standard supplied as a solid for research and analytical applications. The compound is the dodecyl ester of gallic acid (CAS 1166-52-5), chemical formula C19H30O5, molar mass 338.4 g/mol. It is provided with documentation for safe handling and quality verification, and is commonly used in antioxidant, cosmetic, and pharmaceutical research.
Catalog No. 50-002-37964