Learn More
Medchemexpress LLC Dodecyl gallate | 1166-52-5 | MFCD00002195 | 100.0% | 338.44 g/mol | C19H30O5 | 10MM 1ML

Supplier: Medchemexpress LLC HY12408210MM 1ML
Dodecyl gallate is the dodecyl (lauryl) ester of gallic acid used as an antioxidant and research reagent. It is supplied at high purity and is available as solid material or as a pre-made 10 mM solution in DMSO for biological and analytical applications.
- High purity, reported as 99.99%.
- Available as 10 mM in DMSO solution and multiple solid pack sizes (25 g-500 g and larger).
- Molecular weight 338.44 g/mol; chemical formula C19H30O5.
- Commonly used as an antioxidant and antibacterial reagent in research.
- Pre-dissolved solution reduces preparation time and improves dosing accuracy.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.