Learn More
Medchemexpress LLC Dodecylbenzenesulfonic acid | 27176-87-0 | MFCD00066677 | 83.4% | 326.50 g/mol | C18H30O3S | 500 G
SDP
Supplier:  Medchemexpress LLC HYW250144500G
Encompass_Preferred
Dodecylbenzenesulfonic acid is an organosulfonic acid supplied as a liquid and used primarily as a dopant for conductive polymers and as a strong acid catalyst for amino-crosslinked coatings. The material's physical properties and assay are documented on a certificate of analysis for quality control.
  • Acts as a dopant for conductive polymers.
  • Serves as a strong acid catalyst for amino-crosslinked coatings.
  • Provided as a colorless to light yellow liquid with reported density.
  • Purity verified by titration and reported on the COA.
  • Suitable for research and material synthesis applications.
Catalog No. 50-002-80590