Learn More
Abcam Dodecylglucopyranoside, Non-ionic detergent, 100MG
Supplier:  Abcam AB146340100MG
Encompass_Preferred
MW 348.5 g/mol, Purity >99%. Non-ionic detergent for solubilization of membrane-bound proteins in their native state. Long chain alkyl glycopyranoside and a classical nonionic amphiphile surfactant.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4701