Learn More
Medchemexpress LLC Dodecylphosphocholin 50mg | 130890-78-7 | 389.69 g/mol | C17H38NO4P | 50 MG
SDP
Supplier:  Medchemexpress LLC HY116013S50MG
Encompass_Preferred
Dodecyl phosphocholine-d38 is the deuterated form of dodecylphosphocholine (DPC), a zwitterionic detergent used to solubilize membrane proteins for NMR and other biophysical studies. The deuterated (d38) isotopologue reduces proton background, improving spectral quality in solution and solid-state NMR.
  • Deuterated (d38) isotopologue reduces proton background in NMR experiments.
  • Suitable for membrane protein solubilization and structural studies.
  • High purity for analytical and spectroscopic applications.
  • Powder form, typically stored at -20°C to preserve stability.
  • Available in milligram-scale quantities for research use.
Catalog No. 50-002-28260