Learn More
Medchemexpress LLC Dodecyltrimethylammonium bromide | 1119-94-4 | 99.9% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYD0838
Encompass_Preferred
Dodecyltrimethylammonium bromide (DTAB) is a surfactant that interacts with DNA, changing its mechanical properties upon binding and influencing specific binding parameters. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13767