Learn More
Medchemexpress LLC Dodecyltrimethylammonium-d3 chloride | 1219337-20-8 | MFCD28899604 | 266.91 | C15H31D3ClN | 5 MG

Supplier: Medchemexpress LLC HYW7007705MG
DTAC-d3 (dodecyltrimethylammonium-d3 chloride) is the deuterium-labeled analogue of DTAC supplied for use as an isotope-labeled biochemical reagent in life-science research. It is intended for research use only; lot-specific purity and storage instructions are provided in the certificate of analysis.
- Isotopically labeled for mass spectrometry and tracing studies.
- Suitable as an isotope-labeled reagent in biochemical assays.
- Molecular weight 266.91.
- CAS number 1219337-20-8.
- Packaged as 5 mg units.
- For research use only; not for therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.