Learn More
TARGETMOL CHEMICALS INC 7-Methylxanthine 50MG
Supplier: TARGETMOL CHEMICALS INC T054350MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. 7-Methylxanthine is a methyl derivative of xanthine, the purine component of human urinary calculi.7-Methylxanthine has an affinity for adenosine receptors.7-Methylxanthine is a methyl derivative of xanthine, the purine component of human urinary calculi.7-Methylxanthine has an affinity for adenosine receptors. Purity 98.58%