Learn More
Cayman Chemical DofetIlIde N-oxIde 25mg
SDP
Supplier:  Cayman Chemical 3629425 MG
Encompass_Preferred
A metabolite of dofetilide; a less potent antiarrhythmic agent than dofetilide in isolated guinea pig papillary muscle
Catalog No. 50-291-2910